siRNA Details
Gene Symbol: RTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134888.2 | NP_001128360.1 | 1 | 1530 | ||
GenBank mRNA | BC150617.1 | - | 1 | 1530 | 9056987 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134888.2 | NP_001128360.1 | 1 | 1530 | ||
GenBank mRNA | BC150617.1 | - | 1 | 1530 | 9056987 |