siRNA Details
Gene Symbol: LOC79999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291904.1 | NP_001278833.1 | 2, 3 | 252 | ||
XM_017025119.1 | XP_016880608.1 | 3, 4 | 428 | |||
GenBank mRNA | BU595835.1 | - | 2, 3 | 263 |
Gene Symbol: LOC102724624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524126.2 | XP_011522428.1 | 3, 4 | 441 |
Gene Symbol: LOC388436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291905.1 | NP_001278834.1 | 2, 3 | 252 | ||
GenBank mRNA | BC058012.1 | - | 2, 3 | 264 |
