siRNA Details
Gene Symbol: ZNF808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039886.3 | NP_001034975.2 | 5 | 1741 | ||
NM_001321424.1 | NP_001308353.1 | 5 | 1886 | |||
NM_001321425.1 | NP_001308354.1 | 4 | 1639 | |||
XM_005258909.3 | XP_005258966.1 | 4 | 1602 | |||
GenBank mRNA | CR749856.1 | - | 3 | 1517 |
