siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 3 | 178 | ||
XM_011541447.2 | XP_011539749.1 | 4 | 373 | |||
GenBank mRNA | BC130499.1 | - | 3 | 172 | 40146530 | |
BQ435566.1 | - | 4 | 396 | |||
BQ435801.1 | - | 4 | 396 | |||
BU567193.1 | - | 3 | 257 | |||
BU852979.1 | - | 396 |
