siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 4, 5 | 406 | ||
XM_011541447.2 | XP_011539749.1 | 5, 6 | 601 | |||
GenBank mRNA | AA861282.1 | - | 2, 3 | 347 | ||
BC130499.1 | - | 4, 5 | 400 | 40146530 | ||
BC171742.1 | - | 4, 5 | 426 | |||
BQ435801.1 | - | 5, 6 | 624 | |||
BU567193.1 | - | 4, 5 | 485 |
