siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 5 | 461 | ||
XM_011541447.2 | XP_011539749.1 | 6 | 656 | |||
GenBank mRNA | AA861282.1 | - | 2 | 292 | ||
BC130499.1 | - | 5 | 455 | 40146530 | ||
BC171742.1 | - | 5 | 481 | |||
BQ435801.1 | - | 6 | 679 | |||
BU567193.1 | - | 5 | 540 |
