siRNA Details
Gene Symbol: FAM231D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC136705.1 | - | 1 | 542 |
Gene Symbol: LOC100996721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005277507.4 | XP_005277564.1 | 1 | 1469 |
Gene Symbol: LOC388692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123662.1 | - | 1 | 2625 | ||
BC141801.1 | - | 1 | 1385 |
