siRNA Details
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320493.1 | NP_001307422.1 | 3 | 1037 | ||
NM_001320494.1 | NP_001307423.1 | 3 | 878 | |||
NM_016206.3 | NP_057290.2 | 3 | 1037 | |||
XM_006713138.3 | XP_006713201.1 | 3 | 1204 | |||
GenBank mRNA | BC094780.1 | - | 3 | 1033 | 30528902 | |
BX369995.2 | - | 2 | 378 | |||
BX391744.2 | - | 1 | 142 | |||
CR936700.1 | - | 3 | 673 |
