siRNA Details
Gene Symbol: TMEM212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164436.1 | NP_001157908.1 | 1 | 96 | ||
XM_011512817.1 | XP_011511119.1 | 1 | 96 | |||
XM_017006376.1 | XP_016861865.1 | 1 | 106 | |||
GenBank mRNA | AK026825.1 | - | 1 | 76 | ||
DB199311.1 | - | 1 | 96 |
