siRNA Details
Gene Symbol: C5orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145132.1 | NP_001138604.1 | 3 | 527 | ||
XM_011534416.1 | XP_011532718.1 | 4 | 553 | |||
GenBank mRNA | AI187821.1 | - | 1 | 178 | ||
BG721329.1 | - | 3 | 350 |
