siRNA Details
Gene Symbol: LOC107984125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014157.1 | XP_016869646.1 | 16 | 2464 | ||
XM_017014158.1 | XP_016869647.1 | 11 | 1800 |
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304443.1 | - | 23 | 2917 | ||
AK316511.1 | - | 23 | 3351 |
