siRNA Details
Gene Symbol: LOC389906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034031.1 | 4 | 1525 | |||
GenBank mRNA | BC017231.2 | - | 3 | 583 | ||
BC031508.2 | - | 2 | 86 | |||
BC035462.2 | - | 2 | 86 | |||
BC038894.1 | - | 3 | 192 |
