siRNA Details
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321601.1 | NP_001308530.1 | 2 | 212 | ||
GenBank mRNA | BC157867.1 | - | 2 | 226 | ||
BC171868.1 | - | 2 | 226 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321601.1 | NP_001308530.1 | 2 | 212 | ||
GenBank mRNA | BC157867.1 | - | 2 | 226 | ||
BC171868.1 | - | 2 | 226 |