siRNA Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013659.2 | NP_001013681.2 | 7 | 632 | ||
XM_005258927.3 | XP_005258984.1 | 7 | 744 | |||
XM_006723213.3 | XP_006723276.1 | 6 | 654 | |||
GenBank mRNA | AK131417.1 | - | 7 | 632 | ||
AK303740.1 | - | 5 | 425 | |||
BC150303.1 | - | 7 | 694 | |||
BX537524.1 | - | 7 | 676 | |||
DB180171.1 | - | 5 | 427 |
