siRNA Details
Gene Symbol: LOC105369140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003053.1 | XP_016858542.1 | 2 | 730 | ||
XM_017003054.1 | XP_016858543.1 | 2 | 730 | |||
XM_017003055.1 | XP_016858544.1 | 2 | 730 |
Gene Symbol: LOC105369199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542532.2 | XP_011540834.1 | 2 | 1041 | ||
XM_011542534.2 | XP_011540836.1 | 2 | 1043 | |||
XM_017003016.1 | XP_016858505.1 | 2 | 1047 | |||
XM_017003017.1 | XP_016858506.1 | 2 | 1040 | |||
XM_017003018.1 | XP_016858507.1 | 2 | 1045 |
