siRNA Details
Gene Symbol: OR2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012365.1 | NP_036497.1 | 1 | 335 | ||
GenBank mRNA | BC136858.1 | - | 1 | 369 | 9020849 | |
BC136860.1 | - | 1 | 369 | 9020851 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012365.1 | NP_036497.1 | 1 | 335 | ||
GenBank mRNA | BC136858.1 | - | 1 | 369 | 9020849 | |
BC136860.1 | - | 1 | 369 | 9020851 |