siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 1281 | ||
NM_001278662.1 | NP_001265591.1 | 5 | 1261 | |||
NM_001278663.1 | NP_001265592.1 | 3 | 1058 | |||
NM_001278664.1 | NP_001265593.1 | 3 | 1019 | |||
NM_001278677.1 | NP_001265606.1 | 6 | 1353 | |||
NM_001278678.1 | NP_001265607.1 | 2 | 923 | |||
NM_203282.3 | NP_975011.3 | 4 | 1146 | |||
XM_011528443.1 | XP_011526745.1 | 6 | 1414 | |||
XM_011528444.1 | XP_011526746.1 | 7 | 1434 | |||
XM_011528448.2 | XP_011526750.1 | 4 | 1093 | |||
XM_017027514.1 | XP_016883003.1 | 6 | 1344 | |||
XM_017027515.1 | XP_016883004.1 | 8 | 1771 | |||
XM_017027516.1 | XP_016883005.1 | 4 | 3818 | |||
XM_017027517.1 | XP_016883006.1 | 5 | 1647 | |||
XM_017027518.1 | XP_016883007.1 | 4 | 1143 | |||
XM_017027519.1 | XP_016883008.1 | 5 | 1422 | |||
GenBank mRNA | AK291286.1 | - | 4 | 1113 | ||
AK303431.1 | - | 3 | 992 | |||
BC043147.1 | - | 4 | 1118 | 5296940 | ||
BC139749.1 | - | 4 | 1077 | 40115903 | ||
DA852860.1 | - | 1 | 320 |
