siRNA Details
Gene Symbol: LOC283038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033848.1 | 5 | 1061 | |||
GenBank mRNA | BC033536.1 | - | 5 | 1060 | ||
BC048981.1 | - | 5 | 1059 |
Gene Symbol: FLJ37035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033847.1 | 4 | 424 | |||
GenBank mRNA | AK094354.1 | - | 4 | 424 |
