siRNA Details
Gene Symbol: LOC100128108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021852.1 | XP_016877341.1 | 1 | 334 |
Gene Symbol: LOC400464 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127420.1 | - | 1 | 228 |
