siRNA Details
Gene Symbol: LOC400707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124070.1 | - | 1 | 446 | ||
BC132839.1 | - | 1 | 370 | |||
BC132841.1 | - | 1 | 370 |
Gene Symbol: PNMAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020709.2 | NP_065760.1 | 1 | 956 | ||
GenBank mRNA | AK124720.1 | - | 1 | 956 | ||
BX390603.1 | - | 1 | 165 |
