siRNA Details
Gene Symbol: SH2D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103160.1 | NP_001096630.1 | 5, 6 | 1010 | ||
NM_001103161.1 | NP_001096631.1 | 6, 7 | 1139 | |||
XM_011541459.1 | XP_011539761.1 | 6, 7 | 1604 | |||
XM_011541460.1 | XP_011539762.1 | 7, 8 | 1444 | |||
XM_011541461.2 | XP_011539763.1 | 6, 7 | 717 | |||
GenBank mRNA | AK124869.1 | - | 5, 6 | 1010 | ||
AK296431.1 | - | 6, 7 | 708 |
