siRNA Details
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 5 | 1417 | ||
XM_011515604.2 | XP_011513906.1 | 6 | 1603 | |||
XM_011515608.2 | XP_011513910.1 | 6 | 1507 | |||
XM_011515609.2 | XP_011513911.1 | 4 | 1398 | |||
XM_011515610.2 | XP_011513912.1 | 7 | 1684 | |||
XM_011515611.2 | XP_011513913.1 | 5 | 1551 | |||
XM_017012777.1 | XP_016868266.1 | 4 | 1252 | |||
XM_017012778.1 | XP_016868267.1 | 5 | 1455 | |||
GenBank mRNA | AF277624.1 | - | 5 | 1417 |
Gene Symbol: ZNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003441.2 | NP_003432.1 | 4 | 1198 | ||
XM_011513562.2 | XP_011511864.1 | 3 | 2028 | |||
XM_011513563.2 | XP_011511865.1 | 2 | 1786 | |||
XM_017008591.1 | XP_016864080.1 | 4 | 2120 | |||
GenBank mRNA | BC074853.2 | - | 4 | 1058 | 30915571 | |
L15309.1 | - | 4 | 1176 |
Gene Symbol: ZNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256648.1 | NP_001243577.1 | 7 | 1376 | ||
NM_001256649.1 | NP_001243578.1 | 7 | 1241 | |||
NM_001256650.1 | NP_001243579.1 | 6 | 1113 | |||
NM_001256651.1 | NP_001243580.1 | 3 | 727 | |||
NM_001256653.1 | NP_001243582.1 | 4 | 854 | |||
NM_001256654.1 | NP_001243583.1 | 3 | 736 | |||
NM_003423.3 | NP_003414.2 | 4 | 863 | |||
XM_011528257.2 | XP_011526559.1 | 4 | 785 | |||
XM_011528259.2 | XP_011526561.1 | 4 | 1342 | |||
XM_017027207.1 | XP_016882696.1 | 7 | 6854 | |||
XM_017027208.1 | XP_016882697.1 | 7 | 1534 | |||
XM_017027209.1 | XP_016882698.1 | 7 | 6587 | |||
XM_017027210.1 | XP_016882699.1 | 6 | 6378 | |||
XM_017027211.1 | XP_016882700.1 | 7 | 6521 | |||
XM_017027212.1 | XP_016882701.1 | 6 | 1381 | |||
XM_017027213.1 | XP_016882702.1 | 7 | 6629 | |||
XM_017027214.1 | XP_016882703.1 | 5 | 1304 | |||
XM_017027215.1 | XP_016882704.1 | 6 | 1143 | |||
XM_017027216.1 | XP_016882705.1 | 5 | 929 | |||
GenBank mRNA | AF112859.1 | - | 1 | 280 | ||
AK291353.1 | - | 7 | 1191 | |||
BC006528.2 | - | 4 | 830 | 3687640 | ||
BT009756.1 | - | 3 | 675 | |||
CR933640.1 | - | 7 | 1336 | |||
X59244.1 | - | 6 | 1012 |
Gene Symbol: ZNF595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286052.1 | NP_001272981.1 | 3 | 795 | ||
NM_001286053.1 | NP_001272982.1 | 2 | 668 | |||
NM_001286054.1 | NP_001272983.1 | 5 | 992 | |||
NM_182524.3 | NP_872330.1 | 4 | 891 | |||
XM_011513398.2 | XP_011511700.1 | 4 | 1510 | |||
GenBank mRNA | AK056302.1 | - | 4 | 845 | ||
AK300505.1 | - | 3 | 785 | |||
AK301848.1 | - | 2 | 634 | |||
BC036110.1 | - | 4 | 855 | 5297405 | ||
BC043151.1 | - | 4 | 860 | 5297425 | ||
BX537887.1 | - | 4 | 843 |
Gene Symbol: ZNF679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153363.2 | NP_699194.2 | 5 | 1076 | ||
XM_017011797.1 | XP_016867286.1 | 5 | 1037 | |||
GenBank mRNA | BC033523.1 | - | 5 | 1077 | 4821935 |
Gene Symbol: ZNF681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138286.2 | NP_612143.2 | 4 | 1747 | ||
GenBank mRNA | AK056088.1 | - | 3 | 1545 | ||
AK122869.1 | - | 4 | 1751 | |||
BC136346.1 | - | 4 | 1626 | 9020333 |
Gene Symbol: ZNF716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159279.1 | NP_001152751.1 | 4 | 1003 | ||
GenBank mRNA | AK131575.1 | - | 4 | 1003 |
Gene Symbol: ZNF718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039127.5 | NP_001034216.2 | 4 | 1299 | ||
NM_001289930.1 | NP_001276859.1 | 4 | 1922 | |||
NM_001289931.1 | NP_001276860.1 | 4 | 1224 | |||
NR_110529.1 | 2 | 1076 | ||||
XM_017007979.1 | XP_016863468.1 | 5 | 3500 | |||
XM_017007980.1 | XP_016863469.1 | 5 | 3082 | |||
XM_017007981.1 | XP_016863470.1 | 5 | 1329 | |||
GenBank mRNA | AK074517.1 | - | 1 | 708 | ||
AK096662.1 | - | 3 | 1954 | |||
AL833758.1 | - | 2 | 1032 | |||
BC047874.1 | - | 1 | 1234 | |||
BC104026.1 | - | 4 | 1860 | 40025602 | ||
BC104027.2 | - | 4 | 1237 | 40025603 | ||
BC104028.1 | - | 4 | 1860 | 40025604 | ||
BC104029.2 | - | 4 | 1237 | 40025606 | ||
BC154406.1 | - | 4 | 1237 | 40112082 |
Gene Symbol: ZNF721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133474.3 | NP_597731.2 | 3 | 1883 | ||
GenBank mRNA | AB075862.1 | - | 1 | 703 | ||
AK092362.1 | - | 1 | 510 | |||
AK095558.1 | - | 1 | 44 | |||
AK095689.1 | - | 1 | 620 | |||
AK127067.1 | - | 1 | 1439 | |||
AK128499.1 | - | 1 | 1712 | |||
AL834155.1 | - | 1015 |
Gene Symbol: ZNF732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 599 | ||
GenBank mRNA | AK302099.1 | - | 3 | 599 |
Gene Symbol: ZNF733P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003952.1 | 3 | 781 |
Gene Symbol: ZNF735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159524.1 | NP_001152996.1 | 4 | 978 |
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 5 | 1855 | ||
NM_001256173.1 | NP_001243102.1 | 3 | 1638 | |||
NM_003429.4 | NP_003420.2 | 4 | 1765 | |||
XM_011528263.2 | XP_011526565.1 | 6 | 1926 | |||
XM_011528264.1 | XP_011526566.1 | 2 | 2086 | |||
XM_017027234.1 | XP_016882723.1 | 2 | 1565 | |||
GenBank mRNA | AK308258.1 | - | 4 | 1594 | ||
BC047646.1 | - | 2 | 2068 | 4828290 | ||
D70831.1 | - | 1 | 1164 | |||
U35376.1 | - | 4 | 1735 |
Gene Symbol: ZNF876P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027481.1 | 2 | 1057 | |||
GenBank mRNA | BC028359.1 | - | 2 | 1052 | ||
BC046475.1 | - | 2 | 1027 |
