siRNA Details
Gene Symbol: FAM225B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024376.1 | 3 | 5689 | |||
GenBank mRNA | AL045730.1 | - | 1 | 438 |
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125961.1 | - | 1 | 3013 | ||
CR627046.1 | - | 2 | 5879 |
