siRNA Details
Gene Symbol: FAM166A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001710.2 | NP_001001710.1 | 3 | 434 | ||
XM_011518694.1 | XP_011516996.1 | 4 | 1419 | |||
XM_011518696.2 | XP_011516998.1 | 4 | 1419 | |||
XM_011518697.1 | XP_011516999.1 | 4 | 1419 | |||
XM_017014718.1 | XP_016870207.1 | 3 | 1274 | |||
GenBank mRNA | AK097419.1 | - | 3 | 412 | ||
AY604177.1 | - | 3 | 412 | |||
BC132916.1 | - | 3 | 379 | 40146938 | ||
BC132918.1 | - | 3 | 379 | 40146940 |
