siRNA Details
Gene Symbol: FAM224B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002160.1 | 3 | 276 | |||
GenBank mRNA | AI819212.1 | - | 1 | 185 |
Gene Symbol: FAM224A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002161.1 | 3 | 297 | |||
GenBank mRNA | AI670915.1 | - | 1 | 157 | ||
BF476242.1 | - | 1 | 163 |
