siRNA Details
Gene Symbol: ANKRD62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277333.1 | NP_001264262.1 | 1 | 316 | ||
XM_017025720.1 | XP_016881209.1 | 1 | 369 | |||
GenBank mRNA | BX648696.1 | - | 1 | 317 |
