siRNA Details
Gene Symbol: KPNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516215.2 | XP_011514517.1 | 5, 6 | 919 | ||
XM_017012208.1 | XP_016867697.1 | 5, 6 | 919 | |||
XM_017012209.1 | XP_016867698.1 | 5, 6 | 907 | |||
XM_017012210.1 | XP_016867699.1 | 5, 6 | 995 | |||
XM_017012211.1 | XP_016867700.1 | 5, 6 | 919 | |||
XM_017012212.1 | XP_016867701.1 | 5, 6 | 709 | |||
XM_017012213.1 | XP_016867702.1 | 5, 6 | 1047 | |||
XM_017012214.1 | XP_016867703.1 | 5, 6 | 623 | |||
XM_017012215.1 | XP_016867704.1 | 5, 6 | 919 | |||
XM_017012216.1 | XP_016867705.1 | 2, 3 | 367 | |||
GenBank mRNA | EU126604.1 | - | 4, 5 | 563 |
