siRNA Details
Gene Symbol: KPNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516215.2 | XP_011514517.1 | 7 | 1222 | ||
XM_017012208.1 | XP_016867697.1 | 7 | 1222 | |||
XM_017012209.1 | XP_016867698.1 | 7 | 1210 | |||
XM_017012210.1 | XP_016867699.1 | 7 | 1298 | |||
XM_017012211.1 | XP_016867700.1 | 7 | 1222 | |||
XM_017012212.1 | XP_016867701.1 | 7 | 1012 | |||
XM_017012213.1 | XP_016867702.1 | 7 | 1350 | |||
XM_017012214.1 | XP_016867703.1 | 7 | 926 | |||
XM_017012215.1 | XP_016867704.1 | 7 | 1222 | |||
XM_017012216.1 | XP_016867705.1 | 4 | 670 | |||
GenBank mRNA | EU126604.1 | - | 6 | 866 |
