siRNA Details
Gene Symbol: C8orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001795.1 | NP_001001795.1 | 2 | 348 | ||
GenBank mRNA | BC064373.1 | - | 2 | 348 | 5722862 | |
BC064829.1 | - | 2 | 280 | 6176032 | ||
BC073936.1 | - | 2 | 366 | 5223300 |
