siRNA Details
Gene Symbol: SMCO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013698.2 | NP_001013720.2 | 2 | 714 | ||
XM_017019312.1 | XP_016874801.1 | 3 | 3379 | |||
GenBank mRNA | AK092637.1 | - | 2 | 714 | ||
BC132752.1 | - | 2 | 690 | 40146774 | ||
BC132754.1 | - | 2 | 690 | 40146776 | ||
BM696674.1 | - | 1 | 237 |
