siRNA Details
Gene Symbol: CCDC85C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144995.1 | NP_001138467.1 | 1 | 650 | ||
XM_011536705.2 | XP_011535007.1 | 1 | 294 | |||
XM_011536706.2 | XP_011535008.1 | 1 | 294 | |||
XM_011536707.2 | XP_011535009.1 | 1 | 294 |
