siRNA Details
Gene Symbol: OTUD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130901.1 | NP_570971.1 | 1 | 184 | ||
GenBank mRNA | AJ430383.1 | - | 1 | 184 | ||
AK126002.1 | - | 5 | 439 | |||
AK308630.1 | - | 4 | 385 | |||
BC035668.1 | - | 3 | 260 | |||
KJ534968.1 | - | 1 | 91 |
Gene Symbol: LOC100288637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038253.1 | 5 | 541 | |||
NR_038254.1 | 4 | 441 | ||||
NR_038255.1 | 3 | 346 | ||||
GenBank mRNA | AL578899.3 | - | 1 | 57 | ||
BC112921.1 | - | 3 | 218 | |||
BX954662.1 | - | 3 | 324 | |||
BX954797.1 | - | 4 | 442 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_130880, AJ430384.
Gene Ontology Categories:
