siRNA Details
Gene Symbol: LOC100288637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038253.1 | 5, 6 | 594 | |||
NR_038254.1 | 4, 5 | 494 | ||||
NR_038255.1 | 3, 4 | 399 | ||||
GenBank mRNA | BC112921.1 | - | 3, 4 | 271 | ||
BX954662.1 | - | 3, 4 | 377 | |||
BX954797.1 | - | 4, 5 | 495 |
