siRNA Details
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145204.2 | NP_001138676.2 | 1 | 903 | ||
NM_001145205.1 | NP_001138677.2 | 1 | 903 | |||
XM_005255539.3 | XP_005255596.2 | 1 | 904 | |||
XM_011522642.2 | XP_011520944.1 | 1 | 904 | |||
GenBank mRNA | AK299610.1 | - | 2 | 333 | ||
AK309038.1 | - | 1 | 244 | |||
CR627300.1 | - | 1 | 99 | |||
CR749545.1 | - | 1 | 99 | |||
DC375222.1 | - | 2 | 334 |
