siRNA Details
Gene Symbol: LOC105371849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135648.1 | 2 | 402 | |||
GenBank mRNA | BU630466.1 | - | 1 | 307 |
Gene Symbol: WFDC21P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030732.1 | 1 | 66 |
