siRNA Details
Gene Symbol: CLEC4GP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002931.2 | 5, 6 | 531 | |||
GenBank mRNA | AK090689.1 | - | 5, 6 | 531 | ||
AK090734.1 | - | 5, 6 | 528 | |||
AK123140.1 | - | 5, 6 | 512 | |||
BC047593.1 | - | 3, 4 | 420 |
