siRNA Details
Gene Symbol: ZNF729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242680.1 | NP_001229609.1 | 4 | 3100 | ||
XM_017026122.1 | XP_016881611.1 | 5 | 2642 |
Gene Symbol: ZNF99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080409.2 | NP_001073878.2 | 4 | 2989 |
