siRNA Details
Gene Symbol: LINC00665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038278.1 | 1, 2 | 277 | |||
NR_038279.1 | 1, 2 | 277 | ||||
GenBank mRNA | BC041949.1 | - | 1, 2 | 277 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038278.1 | 1, 2 | 277 | |||
NR_038279.1 | 1, 2 | 277 | ||||
GenBank mRNA | BC041949.1 | - | 1, 2 | 277 |