siRNA Details
Gene Symbol: MINOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032363.3 | NP_001027535.1 | 3 | 209 | ||
NM_001204082.1 | NP_001191011.1 | 2 | 161 | |||
NM_001204083.1 | NP_001191012.1 | 3 | 209 | |||
NR_033757.2 | 4 | 394 | ||||
NR_033758.2 | 4 | 288 | ||||
GenBank mRNA | AJ711911.1 | - | 4 | 285 | ||
AK094318.1 | - | 3 | 186 | |||
BC009927.1 | - | 4 | 264 | |||
BC070388.1 | - | 3 | 196 | 30407082 | ||
BG687187.1 | - | 217 | ||||
BM784596.1 | - | 2 | 114 | |||
CB157536.1 | - | 3 | 145 | |||
DA523181.1 | - | 3 | 210 | |||
DA793500.1 | - | 4 | 351 |
