siRNA Details
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034150.1 | 1 | 183 | |||
NR_034151.1 | 1 | 183 | ||||
GenBank mRNA | AK024293.1 | - | 1 | 167 | ||
AK309664.1 | - | 1 | 167 | |||
DA707241.1 | - | 1 | 183 |
