siRNA Details
Gene Symbol: TRIM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004342.3 | NP_001004342.3 | 5 | 1489 | ||
NM_001300889.1 | NP_001287818.1 | 7 | 1303 | |||
XM_011544192.2 | XP_011542494.1 | 5 | 1489 | |||
XM_017001323.1 | XP_016856812.1 | 5 | 1489 | |||
GenBank mRNA | AK126782.1 | - | 5 | 1489 | ||
AY253917.1 | - | 7 | 1261 |
