siRNA Details
Gene Symbol: LINC00895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024381.1 | 1 | 548 | |||
GenBank mRNA | AK057431.1 | - | 1 | 548 | ||
BC042984.2 | - | 1 | 516 | |||
BC104250.1 | - | 1 | 490 | |||
BC104251.1 | - | 1 | 487 | |||
BC113002.1 | - | 1 | 490 | |||
BC113003.1 | - | 1 | 490 |
