siRNA Details
Gene Symbol: LOC107985555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029168.1 | XP_016884657.1 | 4 | 783 | ||
XM_017029169.1 | XP_016884658.1 | 3 | 912 | |||
XM_017029170.1 | XP_016884659.1 | 4 | 348 |
