siRNA Details
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008496.3 | NP_001008496.2 | 5 | 839 | ||
NM_001255975.1 | NP_001242904.1 | 5 | 839 | |||
NR_045648.1 | 6 | 1049 | ||||
NR_045649.1 | 6 | 922 | ||||
GenBank mRNA | AB079368.1 | - | 5 | 835 | ||
AK302409.1 | - | 5 | 839 | |||
AK315806.1 | - | 5 | 839 | |||
BC144693.1 | - | 6 | 679 | |||
BC144696.1 | - | 6 | 806 |
