siRNA Details
Gene Symbol: LOC440982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121655.1 | 1 | 108 | |||
GenBank mRNA | AK054937.1 | - | 1 | 1613 | ||
BX090478.1 | - | 1 | 108 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC440982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121655.1 | 1 | 108 | |||
GenBank mRNA | AK054937.1 | - | 1 | 1613 | ||
BX090478.1 | - | 1 | 108 |