siRNA Details
Gene Symbol: LOC107987363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030106.1 | XP_016885595.1 | 1 | 794 |
Gene Symbol: LOC107986213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008883.1 | XP_016864372.1 | 1 | 1010 |
