siRNA Details
Gene Symbol: LOC646241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134286.1 | 7 | 693 | |||
GenBank mRNA | BC028204.1 | - | 8 | 661 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC646241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134286.1 | 7 | 693 | |||
GenBank mRNA | BC028204.1 | - | 8 | 661 |