siRNA Details
Gene Symbol: C5orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287053.1 | NP_001273982.1 | 5 | 469 | ||
XM_017008942.1 | XP_016864431.1 | 7 | 1589 | |||
GenBank mRNA | BX089473.1 | - | 5 | 467 |
