siRNA Details
Gene Symbol: CCDC192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317938.1 | NP_001304867.1 | 3, 4 | 268 | ||
XM_017009805.1 | XP_016865294.1 | 2, 3 | 330 | |||
XM_017009806.1 | XP_016865295.1 | 3, 4 | 234 | |||
XM_017009807.1 | XP_016865296.1 | 3, 4 | 225 | |||
XM_017009808.1 | XP_016865297.1 | 2, 3 | 156 | |||
XM_017009809.1 | XP_016865298.1 | 3, 4 | 299 | |||
XM_017009810.1 | XP_016865299.1 | 3, 4 | 298 | |||
XM_017009811.1 | XP_016865300.1 | 3, 4 | 298 | |||
XM_017009812.1 | XP_016865301.1 | 3, 4 | 299 | |||
XM_017009813.1 | XP_016865302.1 | 3, 4 | 297 | |||
GenBank mRNA | HY052603.1 | - | 3, 4 | 236 |
