siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 4 | 1785 | |||
GenBank mRNA | AK024177.1 | - | 4 | 1337 | ||
AK092686.1 | - | 3 | 562 | |||
BC036842.1 | - | 2 | 336 | |||
BC038559.1 | - | 4 | 735 | |||
BC044229.1 | - | 2 | 484 | |||
BC112296.1 | - | 1 | 296 | |||
BC112298.1 | - | 1 | 296 |
